Learn More
Cayman Chemical 12-methyl TrIdecnoIc AcId 10mg
SDP
Catalog No. 502912485
Encompass_Preferred
A methylated fatty acid; has been found in bovine milk; topical ocular application decreases angiogenesis and corneal opacity in mouse models of alkali- or P. aeruginosa-induced eye injury at 200 µM
Catalog No. 50-291-2485 Supplier Cayman Chemical Supplier No. 3646110 MG
A methylated fatty acid; has been found in bovine milk; topical ocular application decreases angiogenesis and corneal opacity in mouse models of alkali- or P. aeruginosa-induced eye injury at 200 µM