Learn More
Waters Corp Waters Crimp Cap, Aluminum, PTFE/Silicone/PTFE Septum, 12 mm
Catalog No. 50784215 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These replacement caps are designed for TruView LC/MS Certified Vials. They are reliable for sample storage, offering a long shelf life and protection against leakage. The PTFE/Silicone septum provides effective resealing for repeated injections and sample storage. These caps are compatible with almost all HPLC systems.
  • Chemically inert materials
  • No tools required for assembly
  • Long shelf life
  • Protects from leakage
  • PTFE/Silicone septum for repeated injections and sample storage
  • Septum operating temperature range: -40°C to 200°C
  • Chemically resistant septum until punctured
  • Compatible with almost all HPLC systems
Catalog No. 50-784-215 Supplier Waters Corp Supplier No. PSL404231
These replacement caps are designed for TruView LC/MS Certified Vials. They are reliable for sample storage, offering a long shelf life and protection against leakage. The PTFE/Silicone septum provides effective resealing for repeated injections and sample storage. These caps are compatible with almost all HPLC systems.
  • Chemically inert materials
  • No tools required for assembly
  • Long shelf life
  • Protects from leakage
  • PTFE/Silicone septum for repeated injections and sample storage
  • Septum operating temperature range: -40°C to 200°C
  • Chemically resistant septum until punctured
  • Compatible with almost all HPLC systems