Learn More
Waters Corp WATERS CORPORATION Caps, Aluminum, Crimp, PTFE/Silicone, 100/pk
Catalog No. 50784214 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These aluminum crimp caps are supplied with PTFE/silicone septa, designed for secure sealing in sample preparation applications. Each package contains 100 units, providing a convenient solution for various laboratory needs.
  • Features a crimp closure type.
  • Equipped with an aluminum cap.
  • Designed with dimensions of 12 × 32 mm.
  • Includes a PTFE/silicone seal material.
  • Supplied as a complete cap and septum assembly.
  • Suitable for sample preparation.
  • Comes in packages of 100 units.
  • Compatible with a wide range of laboratory solvents.
  • Withstands high temperatures, suitable for heating or autoclaving.
  • Not supplied sterile, but can be sterilized.
  • Offers a typical shelf life of several years when stored properly.
  • Recommended for single use to ensure reliable and consistent results.
Catalog No. 50-784-214 Supplier Waters Corp Supplier No. PSL404219
These aluminum crimp caps are supplied with PTFE/silicone septa, designed for secure sealing in sample preparation applications. Each package contains 100 units, providing a convenient solution for various laboratory needs.
  • Features a crimp closure type.
  • Equipped with an aluminum cap.
  • Designed with dimensions of 12 × 32 mm.
  • Includes a PTFE/silicone seal material.
  • Supplied as a complete cap and septum assembly.
  • Suitable for sample preparation.
  • Comes in packages of 100 units.
  • Compatible with a wide range of laboratory solvents.
  • Withstands high temperatures, suitable for heating or autoclaving.
  • Not supplied sterile, but can be sterilized.
  • Offers a typical shelf life of several years when stored properly.
  • Recommended for single use to ensure reliable and consistent results.