Learn More
Analytical Sales & Services 12 Month Canary-Safe Exhaust Filter
Catalog No. 50646465
Encompass
12 Month, Canary-SafeIDX Vapor Exhaust Filter; Charcoal, 100g; with Expiration Time Strip
Catalog No. 50-646-465 Supplier Analytical Sales & Services Supplier No. CX12000
12 Month, Canary-SafeIDX Vapor Exhaust Filter; Charcoal, 100g; with Expiration Time Strip