Learn More
Cayman Chemical 12-O-Methylcarnosic Acid, Purity: ≥98%, CAS: 62201-71-2, 10 mg
SDP
Catalog No. 502913096
Encompass_Preferred
A diterpene with antileishmanial and gastroprotective activities; induces mortality in L. donovani amastigotes (IC50 = 0.75 µM); reduces lesion area in a mouse model of hydrochloric acid and ethanol-induced gastric ulcers at 10 mg/kg
Catalog No. 50-291-3096 Supplier Cayman Chemical Supplier No. 3617510 MG
A diterpene with antileishmanial and gastroprotective activities; induces mortality in L. donovani amastigotes (IC50 = 0.75 µM); reduces lesion area in a mouse model of hydrochloric acid and ethanol-induced gastric ulcers at 10 mg/kg