Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 12-O-Methylcarnosic acid | 62201-71-2 | 10 MG
SDP

Catalog No. 5000417267
Encompass_Preferred

12-O-Methylcarnosic acid is a diterpene carnosic acid isolated from the acetone extract of Salvia microphylla. It acts as an active constituent inhibiting 5α-reductase with an IC50 value of 61.7 μM. This compound also inhibits proliferation in LNCaP cells and exhibits antioxidant, anti-cancer, and antimicrobial activities.

  • Isolated from the acetone extract of Salvia microphylla.
  • Inhibits 5α-reductase with an IC50 value of 61.7 μM.
  • Inhibits proliferation in LNCaP cells.
  • Possesses antioxidant, anti-cancer, and antimicrobial activities.
  • Purity: 99.67%.
  • Molecular weight: 346.46.
  • Formula: C21H30O4.
  • Appearance: Solid, white to yellow.

Catalog No. 50-004-17267 Supplier Medchemexpress LLC Supplier No. HYN751010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

12-O-Methylcarnosic acid is a diterpene carnosic acid isolated from the acetone extract of Salvia microphylla. It acts as an active constituent inhibiting 5α-reductase with an IC50 value of 61.7 μM. This compound also inhibits proliferation in LNCaP cells and exhibits antioxidant, anti-cancer, and antimicrobial activities.

  • Isolated from the acetone extract of Salvia microphylla.
  • Inhibits 5α-reductase with an IC50 value of 61.7 μM.
  • Inhibits proliferation in LNCaP cells.
  • Possesses antioxidant, anti-cancer, and antimicrobial activities.
  • Purity: 99.67%.
  • Molecular weight: 346.46.
  • Formula: C21H30O4.
  • Appearance: Solid, white to yellow.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.