Learn More
Cayman Chemical 12-OAHSA-d17 100ug
SDP
Catalog No. 502965104
Encompass_Preferred
An internal standard for the quantification of 12-OAHSA by GC- or LC-MS
Catalog No. 50-296-5104 Supplier Cayman Chemical Supplier No. 17195100 UG
An internal standard for the quantification of 12-OAHSA by GC- or LC-MS