Learn More
Cayman Chemical 12-oxo LeukotrIen B4 100ug
SDP
Catalog No. 502982900
Encompass_Preferred
An initial metabolite of LTB4 with significantly reduced biological activity
Catalog No. 50-298-2900 Supplier Cayman Chemical Supplier No. 20140100 UG
An initial metabolite of LTB4 with significantly reduced biological activity