Learn More
Cayman Chemical 12-oxo StearIc AcId 100mg
SDP
Catalog No. 502942145
Encompass_Preferred
A metabolite of ricinoleic acid; has been found in cow and goat milk; dietary administration induces encephalomalacia in chicks fed a vitamin E-deficient diet
Catalog No. 50-294-2145 Supplier Cayman Chemical Supplier No. 40259100 MG
A metabolite of ricinoleic acid; has been found in cow and goat milk; dietary administration induces encephalomalacia in chicks fed a vitamin E-deficient diet