Learn More
Cayman Chemical 12-oxo Stearic Acid, Purity: ≥95%, CAS: 925-44-0, 25 mg
SDP
Catalog No. 502942187
Encompass_Preferred
A metabolite of ricinoleic acid; has been found in cow and goat milk; dietary administration induces encephalomalacia in chicks fed a vitamin E-deficient diet
Catalog No. 50-294-2187 Supplier Cayman Chemical Supplier No. 4025925 MG
A metabolite of ricinoleic acid; has been found in cow and goat milk; dietary administration induces encephalomalacia in chicks fed a vitamin E-deficient diet