Learn More
Cliawaived Inc IDTC-II 12 Instant Drug Test Cards
SDP
Catalog No. NC1542640
Encompass_Preferred
(25/Pack) CLIAwaived, Inc. IDTC-II Instant Drug Test Card - Tests for AMP500 / BAR300 / BUP10 / BZO300 / COC150 / MDMA500 / MET500 / MTD300 / OPI300 / OXY100 / PCP25 / THC50 + Adulterants+ CRE, pH, OX. The newly designed IDTC-II dip card features a hermetic seal to prevent the card from flooding. The device features a flat and separate cap so it can be easily photographed without contamination.
This item is not returnable. View return policy
Catalog No. NC1542640 Supplier Cliawaived Inc Supplier No. CLIAIDTCII88A
(25/Pack) CLIAwaived, Inc. IDTC-II Instant Drug Test Card - Tests for AMP500 / BAR300 / BUP10 / BZO300 / COC150 / MDMA500 / MET500 / MTD300 / OPI300 / OXY100 / PCP25 / THC50 + Adulterants+ CRE, pH, OX. The newly designed IDTC-II dip card features a hermetic seal to prevent the card from flooding. The device features a flat and separate cap so it can be easily photographed without contamination.