Learn More
JANT PHARMACAL CORPORATION Accutest ValuPak Drug Test Cup 12 AMP/BAR/BUP/BZO/COC/MDMA/MET /MOP /MTD/OXY/PCP/THC 25 cups/kit box
SDP
Catalog No. NC3457089
Encompass_Preferred
Accutest CLIA Waived 12 panel drug test cup is a rapid, one-step qualitative urine drug test that uses a visually read competitive binding immunoassay method for screening multiple drugs and drug metabolites in human urine without the need of instrumentation.
Catalog No. NC3457089 Supplier JANT PHARMACAL CORPORATION Supplier No. DS412
Accutest CLIA Waived 12 panel drug test cup is a rapid, one-step qualitative urine drug test that uses a visually read competitive binding immunoassay method for screening multiple drugs and drug metabolites in human urine without the need of instrumentation.