Learn More
Cliawaived Inc CLIAwaived Inc. Instant Drug Test Cup(IDTC) - 12 Panel Cup. Tests for: THC,COC,OPI 300, AMP,mAMP,PCP,BAR,BZO,MTD,OXY,TCA,XTC + adulterants (Box of 25 cups)
SDP
Catalog No. NC2579466
Encompass_Preferred
CLIAwaived, Inc. - INSTANT DRUG TEST CUP (IDTC II). This 12-panel, no-leak drug test cup tests for AMP, BAR, BZO, COC, MDMA, MET, MTD, OPI (MOR 300), OXY, PCP, TCA, and THC + Adulteration (CRE/OX/pH). Results in 5 minutes. FDA-cleared and CLIA-waived.
This item is not returnable. View return policy
Catalog No. NC2579466 Supplier Cliawaived Inc Supplier No. CLIAIDTC12A
CLIAwaived, Inc. - INSTANT DRUG TEST CUP (IDTC II). This 12-panel, no-leak drug test cup tests for AMP, BAR, BZO, COC, MDMA, MET, MTD, OPI (MOR 300), OXY, PCP, TCA, and THC + Adulteration (CRE/OX/pH). Results in 5 minutes. FDA-cleared and CLIA-waived.