Learn More
NEXSCREEN LLC 13 Panel Nexscreen Cup,25/Pk
Supplier:  NEXSCREEN LLC ND60135N
Encompass_Preferred
NexScreen Cup, integrated rapid drug test cup, set the standard for advanced rapid drug testing. It is accurate, easy, rugged, and flexible. Accurate: The NexScreen Cup (FDA-cleared, CLIA-waived, CE marked) provides the same accuracy as a laboratory. In an independent study, the NexScreen Cup provided 100% correlation at 25% and 50% above and below cutoff. NexScreen Cup¿s testing accuracy has been published in the Journal of Analytical Toxicology and endorsed by the Society of Forensic Toxicologists. Simplicity: Just collect the specimen, peel away the label, and read the color-coded results. There¿s only 1 drug test on each strip making it easy to read.
Catalog No. NC1595115