Learn More
Cliawaived Inc 12 Panel Urine Drug Test Card with Buprenorhine
SDP
Catalog No. NC1112620
Encompass_Preferred
(25/Pack) CLIAwaived, Inc. 12 Panel Urine Drug Test Card. Tests for AMP500 / BAR300 / BUP10 / BZO300 / COC150 / MDMA500 / MET500 / MTD300 / OPI (MOR 300) / OXY100 / PCP25 / THC50.The newly designed IDTC-II dip card features a hermetic seal to prevent the card from flooding. The device features a flat and separate cap so it can be easily photographed without contamination.
Catalog No. NC1112620 Supplier Cliawaived Inc Supplier No. CLIAIDTCII88
(25/Pack) CLIAwaived, Inc. 12 Panel Urine Drug Test Card. Tests for AMP500 / BAR300 / BUP10 / BZO300 / COC150 / MDMA500 / MET500 / MTD300 / OPI (MOR 300) / OXY100 / PCP25 / THC50.The newly designed IDTC-II dip card features a hermetic seal to prevent the card from flooding. The device features a flat and separate cap so it can be easily photographed without contamination.