Learn More
BTNX Inc 12 Parameter Urine Drug Screen w/ adultarants - 25/kit (12.11) - MOP300 OXY100 BZO300 OPI2000 MET1000 AMP1000 COC300 THC50 MDMA500 BUP10 BAR300 MTD300 + Cre/pH/Oxi
Catalog No. NC3361253
Encompass_Preferred
The Rapid Response™ Multi-Drug One Step Cup is an easy-to-use, one-step solution for drug screening that can simultaneously detect multiple drugs in urine samples. This format is a great alternative to the industry's complicated testing processes and is available with and without adulterant tests. It has a longer detection window than saliva tests and provides results in 5 minutes. This product is also available with an option of built-in Specimen Validity/Adulteration Test Strips. (12.11) - MOP300 OXY100 BZO300 OPI2000 MET1000 AMP1000 COC300 THC50 MDMA500 BUP10 BAR300 MTD300 + Cre/pH/Oxi
This item is not returnable. View return policy
Catalog No. NC3361253 Supplier BTNX Inc Supplier No. D12.111VA
The Rapid Response™ Multi-Drug One Step Cup is an easy-to-use, one-step solution for drug screening that can simultaneously detect multiple drugs in urine samples. This format is a great alternative to the industry's complicated testing processes and is available with and without adulterant tests. It has a longer detection window than saliva tests and provides results in 5 minutes. This product is also available with an option of built-in Specimen Validity/Adulteration Test Strips. (12.11) - MOP300 OXY100 BZO300 OPI2000 MET1000 AMP1000 COC300 THC50 MDMA500 BUP10 BAR300 MTD300 + Cre/pH/Oxi