Learn More
BTNX Inc 12 Parameter Urine Drug Screen w/ adultarants - 25/kit (12.26) - AMP1000, BUP10, BZO300, COC300, EDDP100, ETG500, FYL10, MDMA500, MET1000, MOP300, OXY100, THC50 + Cre/SG/pH
Catalog No. NC3361255
Encompass_Preferred
The Rapid Response™ Multi-Drug One Step Cup is an easy-to-use, one-step solution for drug screening that can simultaneously detect multiple drugs in urine samples. This format is a great alternative to the industry's complicated testing processes and is available with and without adulterant tests. It has a longer detection window than saliva tests and provides results in 5 minutes. This product is also available with an option of built-in Specimen Validity/Adulteration Test Strips.  (12.26) - AMP1000, BUP10, BZO300, COC300, EDDP100, ETG500, FYL10, MDMA500, MET1000, MOP300, OXY100, THC50 + Cre/SG/pH
This item is not returnable. View return policy
Catalog No. NC3361255 Supplier BTNX Inc Supplier No. D12.261VA
The Rapid Response™ Multi-Drug One Step Cup is an easy-to-use, one-step solution for drug screening that can simultaneously detect multiple drugs in urine samples. This format is a great alternative to the industry's complicated testing processes and is available with and without adulterant tests. It has a longer detection window than saliva tests and provides results in 5 minutes. This product is also available with an option of built-in Specimen Validity/Adulteration Test Strips.  (12.26) - AMP1000, BUP10, BZO300, COC300, EDDP100, ETG500, FYL10, MDMA500, MET1000, MOP300, OXY100, THC50 + Cre/SG/pH