Learn More
BTNX Inc 12 Parameter Urine Drug Screen - 25/kit (12.5) - AMP1000 BAR300 BUP10 BZO300 COC300 MDMA500 MTD300 MET1000 OPI2000 OXY100 THC50 PCP25
Catalog No. NC3361250
Encompass_Preferred
The Rapid Response™ Multi-Drug One Step Cup is an easy-to-use, one-step solution for drug screening that can simultaneously detect multiple drugs in urine samples. This format is a great alternative to the industry's complicated testing processes and is available with and without adulterant tests. It has a longer detection window than saliva tests and provides results in 5 minutes. This product is also available with an option of built-in Specimen Validity/Adulteration Test Strips.  (12.5) - AMP1000 BAR300 BUP10 BZO300 COC300 MDMA500 MTD300 MET1000 OPI2000 OXY100 THC50 PCP25
This item is not returnable. View return policy
Catalog No. NC3361250 Supplier BTNX Inc Supplier No. D12.51V
The Rapid Response™ Multi-Drug One Step Cup is an easy-to-use, one-step solution for drug screening that can simultaneously detect multiple drugs in urine samples. This format is a great alternative to the industry's complicated testing processes and is available with and without adulterant tests. It has a longer detection window than saliva tests and provides results in 5 minutes. This product is also available with an option of built-in Specimen Validity/Adulteration Test Strips.  (12.5) - AMP1000 BAR300 BUP10 BZO300 COC300 MDMA500 MTD300 MET1000 OPI2000 OXY100 THC50 PCP25