Learn More
Thermo Scientific™ 12-Place Swinging Bucket Microliter Rotor with Cover
Shop All Thermo Scientific ProductsDescription
- High RCF of 16,438 x g
- Separation of liquid multi-phase systems
- Centrifugation of filtration units
- Density gradient-separation of cells/cell organelles
- Semi-preparative centrifugation of proteins
Specifications
Specifications
| Type | Swinging Bucket Microliter Rotor |
| Capacity | 12 x 1.5/2 mL |
| Max. RCF | 16,438 x g |
| Max. Speed | 13,000 rpm |
| For Use With (Equipment) | Thermo Scientific Sorvall Primo R Centrifuge |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.