Learn More
USA Scientific Inc 12-box (2 x 3) upright freezer rack for 50-place boxes
Catalog No. 507541183
Encompass
12-box (2 long x 3 high) upright freezer rack for 50-place boxes, 12 3/4 in L x 5 7/8 in W x 7 in H, stainless steel
Catalog No. 50-754-1183 Supplier USA Scientific Inc Supplier No. 26861215
12-box (2 long x 3 high) upright freezer rack for 50-place boxes, 12 3/4 in L x 5 7/8 in W x 7 in H, stainless steel