Learn More
Electron Microscopy Sciences 12" Polypropylene Retriever
SDP
Catalog No. 5029136
Encompass_Preferred
Polypropylene stirring bar retrievers are ideal for use with all stirring bars. The entire retriever is totally encased in a chemically resistant polypropylene, with a handy ring for hanging when not in use.
Catalog No. 50-291-36 Supplier Electron Microscopy Sciences Supplier No. 6420812
Polypropylene stirring bar retrievers are ideal for use with all stirring bars. The entire retriever is totally encased in a chemically resistant polypropylene, with a handy ring for hanging when not in use.