Learn More
Medchemexpress LLC HY-Y0921 100mL Medchemexpress, (±)-1,2-Propanediol CAS:57-55-6 Purity:>98%
SDP
Catalog No. 502083553
Encompass_Preferred
Medchemexpress, HY-Y0921 100mL (±)-1,2-Propanediol CAS:57-55-6 (±)-1,2-Propanediol is an aliphatic alcohol and frequently used as an excipient in many drug formulations to increase the solubility and stability of drugs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3553 Supplier Medchemexpress LLC Supplier No. HYY0921100ML
Medchemexpress, HY-Y0921 100mL (±)-1,2-Propanediol CAS:57-55-6 (±)-1,2-Propanediol is an aliphatic alcohol and frequently used as an excipient in many drug formulations to increase the solubility and stability of drugs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.