Learn More
Sigma Aldrich Fine Chemicals Biosciences 12-Propanediol ReagentPlus1KG
Catalog No. 502898238
Encompass
1 2-Propanediol can be obtained as a byproduct in the dehydration of glycerol to hydroxyacetone.It can undergo condensation with sebacic acid and citric acid in the absence of catalyst to give poly((1 2-propanediol-sebacate)-citrate) (PPSC) an elastomeric polyester which can find applications in tissue engineering and drug delivery.
Catalog No. 50-289-8238 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1343681KG
1 2-Propanediol can be obtained as a byproduct in the dehydration of glycerol to hydroxyacetone.It can undergo condensation with sebacic acid and citric acid in the absence of catalyst to give poly((1 2-propanediol-sebacate)-citrate) (PPSC) an elastomeric polyester which can find applications in tissue engineering and drug delivery.