Learn More
Enzo Life Sciences 12(S),20-Dihydroxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (DiHETE) (50µg). CAS: 89614-44-8
SDP
Supplier:  Enzo Life Sciences BMLD1220050
Encompass_Preferred
Transcellular biosynthesis via interaction between platelets and neutrophils. Formulation: Oil dissolved in ethanol. Purity: ≥99% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-0499