Learn More
Cayman Chemical 12(S)-HETE, 54397-83-0, 25 µg
SDP
Supplier:  Cayman Chemical 3457025
Encompass_Preferred
**Custom size that requires a quote to purchase. Contact Fisher Customer Service to request a quote for current pricing** The predominant LO product of arachidonic acid formed in mammalian platelets; enhances tumor cell adhesion to endothelial cells, fibronectin, and the subendothelial matrix at 0.1 µM
Catalog No. NC0011005