Learn More
Cayman Chemical 12(S)-HETE-d8, 84807-90-9, 25 µg
SDP
Supplier:  Cayman Chemical 33457025
Encompass_Preferred
12(S)-HETE-d8 contains eight deuterium atoms at the 5, 6, 8, 9, 11, 12, 14 and 15 positions. It is intended for use as an internal standard for the quantification of 12(S)-HETE by GC- or LC-mass spectrometry (MS).
Catalog No. NC0679209