Learn More
Ace Glass, Inc. 12 SECTION SNYDER COLUMN 12 SECTION SNYDER COLUMN
SDP
Catalog No. 50978832
Encompass_Preferred
12-section Snyder column for use in distillation applications. Column is floating ball type with 24/40 joints. Each section is approximately 51mm long.
Catalog No. 50-978-832 Supplier Ace Glass, Inc. Supplier No. 657506
12-section Snyder column for use in distillation applications. Column is floating ball type with 24/40 joints. Each section is approximately 51mm long.