Learn More
Bioworld SnapStrip® PCR Tubes w/ Unattached Caps, Assorted, 1 PK(s)
Catalog No. 50197148
Encompass_Preferred
Product Specifications:12-tube strips, 125 total strips per packUnattached dome caps0.2 mL measurementTubes can be easily separated when desired to create smaller stripsAssorted Colors (blue, green, pink, purple, yellow)
Catalog No. 50-197-148 Supplier Bioworld Supplier No. 111900541
Product Specifications:12-tube strips, 125 total strips per packUnattached dome caps0.2 mL measurementTubes can be easily separated when desired to create smaller stripsAssorted Colors (blue, green, pink, purple, yellow)