Learn More
Cayman Chemical 12-Tridecenoic Acid, Purity: ≥95%, CAS: 6006-06-0, 10 mg
SDP
Catalog No. 502917916
Encompass_Preferred
A monounsaturated fatty acid; active against S. aureus (MIC = 1.17 mM); inhibits gastric juice secretion in a rat model of pylorus ligation-induced gastric ulcers at 5, 10, and 25 mg/kg
Catalog No. 50-291-7916 Supplier Cayman Chemical Supplier No. 3760110 MG
A monounsaturated fatty acid; active against S. aureus (MIC = 1.17 mM); inhibits gastric juice secretion in a rat model of pylorus ligation-induced gastric ulcers at 5, 10, and 25 mg/kg