Learn More
Cell Signaling Technology 12-Tube Magnetic Separation Rack, Neodymium Magnets, 1.5 mL to 2.0 mL Tubes, 1 ea
SDP
Catalog No. 502045844
Encompass
The 12-Tube Magnetic Separation Rack is designed for quick and easy small-scale isolation of immunocomplexes using magnetic beads. This rack holds up to twelve 1.5 to 2.0 mL tubes and features six neodymium rare earth permanent magnets to facilitate efficient bead collection during immunoprecipitation and chromatin immunoprecipitation (ChIP) protocols.
  • Holds up to twelve 1.5 to 2.0 mL tubes.
  • Contains six powerful neodymium rare earth permanent magnets.
  • Compatible with common magnetic bead-based immunoprecipitation and chromatin IP kits.
  • Simplified procedure for supernatant removal and bead resuspension.
  • Caution: Device contains powerful magnets that may damage sensitive mechanical or electrical instruments.
Catalog No. 50-204-5844 Supplier Cell Signaling Technology Supplier No. 14654S
The 12-Tube Magnetic Separation Rack is designed for quick and easy small-scale isolation of immunocomplexes using magnetic beads. This rack holds up to twelve 1.5 to 2.0 mL tubes and features six neodymium rare earth permanent magnets to facilitate efficient bead collection during immunoprecipitation and chromatin immunoprecipitation (ChIP) protocols.
  • Holds up to twelve 1.5 to 2.0 mL tubes.
  • Contains six powerful neodymium rare earth permanent magnets.
  • Compatible with common magnetic bead-based immunoprecipitation and chromatin IP kits.
  • Simplified procedure for supernatant removal and bead resuspension.
  • Caution: Device contains powerful magnets that may damage sensitive mechanical or electrical instruments.