Learn More
Alkali Scientific Low Adhesion Cell Culture Plates for Suspension Cultures, 12-Well, Flat, Sterile, 1/pk, 50/cs
SDP
Catalog No. 502335980
Encompass_Preferred
Low Adherence Cell Culture Plates­­­­­­­­­­­­­­­­­­­­­­­­­­­ are manufactured in Class 100,000 grade clean room with USP 6 grade raw materials for optimal performance. Suspension cell growth and proliferation in media (not on a surface) can be expected within these low adherence / low adhesion plates. The plates have an overall neutral charge which minimizes cell spreading. A moderately hydrophilic surface for Cell Cultures that effectively inhibit cell adhesion. Sterilized by E-beam. Peel-to-open, individually wrapped. Non-Pyrogenic, DNase and RNase Free.
Catalog No. 50-233-5980 Supplier Alkali Scientific Supplier No. TL42
Add to Cart
Add to Cart
Low Adherence Cell Culture Plates­­­­­­­­­­­­­­­­­­­­­­­­­­­ are manufactured in Class 100,000 grade clean room with USP 6 grade raw materials for optimal performance. Suspension cell growth and proliferation in media (not on a surface) can be expected within these low adherence / low adhesion plates. The plates have an overall neutral charge which minimizes cell spreading. A moderately hydrophilic surface for Cell Cultures that effectively inhibit cell adhesion. Sterilized by E-beam. Peel-to-open, individually wrapped. Non-Pyrogenic, DNase and RNase Free.