Learn More
Chemglass Life Sciences 12-Well Cell Culture Plate, Non-Treated, Sterile, Flat Bottom Wells
SDP
Catalog No. 501897615
Encompass
• High clarity, 100% virgin polystyrene• Sterilized by E-beam• Vacuum plasma tissue culture treatment• Clear lot number for batch traceability• Peel-to-open, individually wrapped• Markings of well coordinates available for 12, 24 and 96 well plates• Non-pyrogenic, DNase- and RNase-free
Catalog No. 50-189-7615 Supplier Chemglass Life Sciences Supplier No. CGE3501012
Add to Cart
Add to Cart
• High clarity, 100% virgin polystyrene• Sterilized by E-beam• Vacuum plasma tissue culture treatment• Clear lot number for batch traceability• Peel-to-open, individually wrapped• Markings of well coordinates available for 12, 24 and 96 well plates• Non-pyrogenic, DNase- and RNase-free