Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ BIO-CERT™ CELL CULTURE QUALITY 2-in-1 Culture Inserts for 12-well Plates, 8 μL Pore Size, PC Membrane

Catalog No. 14381009 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Packaging:
Multi-Pack Blister, 36/Pk.
Single Blister, 48/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
14-381-009 Multi-Pack Blister, 36/Pk.
14-381-008 Single Blister, 48/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-381-009 Supplier BRANDTECH™ BRAND™ Supplier No. 782727
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sterile inserts for 12-well plates, featuring cellGrade™ plus surface treatment for uniform cell growth. Designed with feet for standing or a removable hook for hanging, offering versatile use in cell culture applications.

  • Suitable for 2D and 3D cell cultures
  • TC treated with cellGrade™ plus surface treatment for enhanced cell growth
  • Translucent PC membrane ideal for transport studies
  • Sterile according to ISO 11137, SAL 10-6
  • Produced in a Class 8 cleanroom per ISO 14644-1
  • Non-cytotoxic, meeting ISO 10993-5 standards
  • BIO-CERT™ CELL CULTURE QUALITY means inserts are free of pyrogens, endotoxins, human DNA, RNase, and DNase
  • Diameter size of 13.3 mm for an ideal fit of 12-well plates
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Breakapart No
Certifications/Compliance Sterile according to ISO 11137, SAL 10-6, Non-cytotoxic according to ISO 10993-5, pyrogen and endotoxin free to 0.01EU/mL
Certified Yes
Lid Without Lid
Content And Storage Ambient
Culture Area 1.38 cm2
Membrane PC
Array 1 x 12
Autoclavable Not Autoclavable
Packaging Multi-Pack Blister, 36/Pk.
Pore Size 8 μm
Binding Type cellGrade plus
Color Clear
Disposable Single-use
Product Type Microplate Insert
For Use With (Equipment) 12-well plate
Material Polystyrene
Volume (Metric) Well 1000 μL
No. of Wells 12
Volume (Metric) Working 300 to 1000 μL
Sterility Sterile
Surface Treatment cellGrade Plus
Well Shape Round, Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) 2D and 3D cell culture, co-culture, migration, chemotaxis, TEER assays, and air-lift culture
Quantity 36
RNase Free Yes
Well Color Clear
Height (Metric) 11 mm
Diameter (Metric) Membrane 13.3 mm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.