Learn More
Altenloh Brinck & Co US INC. DBA: SABEU 12 well insert 0.4 um PET PK48 (cellQART)
Catalog No. 502673512
Encompass_Preferred
These cellQART Cell Culture Inserts come with PET membranes in translucent optics in 12-Well format. The Inserts are tissue culture treated, sterile and are compatible with most standard Cell Culture Plates. They are ideal for fast cell growth / barrier formation.
Catalog No. 50-267-3512 Supplier Altenloh Brinck & Co US INC. Supplier No. 9310402
These cellQART Cell Culture Inserts come with PET membranes in translucent optics in 12-Well format. The Inserts are tissue culture treated, sterile and are compatible with most standard Cell Culture Plates. They are ideal for fast cell growth / barrier formation.