Learn More
Altenloh Brinck & Co US INC. DBA: SABEU 12 well insert 0.4 um PET PK48 (cellQART)
Supplier:  Altenloh Brinck & Co US INC. 9310422
Encompass_Preferred
These cellQART Cell Culture Inserts come with PET membranes in clear extended culture optics in 12-Well format. The Inserts are tissue culture treated, sterile and are compatible with most standard Cell Culture Plates. They are ideal for in-situ phase contrast microscopy. Longer studies/testing (2weeks+) and imaging clarity is best suited for the Clear Extended insert.
Catalog No. 50-267-3508