Learn More
Altenloh Brinck & Co US INC. DBA: SABEU 12 well insert 3.0um PET PK48 (cellQART)
Supplier:  Altenloh Brinck & Co US INC. 9313002
Encompass_Preferred
these cellQART Cell Culture Inserts come with PET membranes in translucent optics in 12-Well format. The Inserts are tissue culture treated, sterile and are compatible with most standard Cell Culture Plates. They are ideal for fast cell growth / barrier formation, cell invasion, cell motility and chemotaxis.
Catalog No. 50-267-3502