Learn More
Altenloh Brinck & Co US INC. DBA: SABEU 12 well insert 8.0um PET PK48 (cellQART)
Catalog No. 502673497
Encompass_Preferred
These cellQART Cell Culture Inserts come with PET membranes in clear optics in 12-Well format. The Inserts are tissue culture treated, sterile and are compatible with most standard Cell Culture Plates. They are ideal for in-situ phase contrast microscopy.
Catalog No. 50-267-3497 Supplier Altenloh Brinck & Co US INC. Supplier No. 9318012
These cellQART Cell Culture Inserts come with PET membranes in clear optics in 12-Well format. The Inserts are tissue culture treated, sterile and are compatible with most standard Cell Culture Plates. They are ideal for in-situ phase contrast microscopy.