Learn More
Quality Biological Inc 12 Well Non-treated Plate with Lid, Individual, Sterile, 100/CS
SDP
Catalog No. 50147460
Encompass_Preferred
12 Well Non-treated Plate, Polystyrene, with Lid, Sterile
Packaged Individually, 50/PK, 100/CS
• Plates are labeled with alpha-numeric markings for easy identification of individual wells
• Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination
• Lids have a stacking feature for added stability when handling tall stacks of plates
• Working volume: 2 - 4 mL
Catalog No. 50-147-460 Supplier Quality Biological Inc Supplier No. P886229512
12 Well Non-treated Plate, Polystyrene, with Lid, Sterile
Packaged Individually, 50/PK, 100/CS
• Plates are labeled with alpha-numeric markings for easy identification of individual wells
• Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination
• Lids have a stacking feature for added stability when handling tall stacks of plates
• Working volume: 2 - 4 mL