Learn More
Bellco Glass, Inc. 12 Well Non-treated Plate with
SDP
Catalog No. 502330382
Encompass_Preferred
DescriptionPlates are labeled with alpha-numeric markings for easy identification of individual wellsEach well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contaminationLids have a stacking feature for added stability when handling tall stacks of plates Pack DescriptionIndividual, 50/PackCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneWorking Volume2–4mLBottom ShapeFlatTissue Culture TreatedNoRNase/DNase FreeYes
Catalog No. 50-233-0382 Supplier Bellco Glass, Inc. Supplier No. 229512000
Add to Cart
Add to Cart
DescriptionPlates are labeled with alpha-numeric markings for easy identification of individual wellsEach well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contaminationLids have a stacking feature for added stability when handling tall stacks of plates Pack DescriptionIndividual, 50/PackCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneWorking Volume2–4mLBottom ShapeFlatTissue Culture TreatedNoRNase/DNase FreeYes