Learn More
MSC Quadrant EPP Nylon Sheet, 3/32 in, 12x12 in, 1 ea
Catalog No. NC1131541
Encompass
This nylon 101 sheet is the strongest and most rigid of unmodified nylons, with one of the highest melting points. It is commonly specified for screw machined electrical insulators and food contact parts, suitable for a temperature range of -40°F to 225°F. It is also compliant with USDA, NSF, and 3A-Dairy standards, and made from FDA-compliant resins.
  • Not UV stabilized
  • Temperature range: -40°F to 225°F
  • FDA-compliant resin/materials
  • NSF/ANSI/CAN listed
  • Meets USDA and 3-A standards
  • Cut tolerance ±1/4 in
Catalog No. NC1131541 Supplier MSC Supplier No. 63396790
This nylon 101 sheet is the strongest and most rigid of unmodified nylons, with one of the highest melting points. It is commonly specified for screw machined electrical insulators and food contact parts, suitable for a temperature range of -40°F to 225°F. It is also compliant with USDA, NSF, and 3A-Dairy standards, and made from FDA-compliant resins.
  • Not UV stabilized
  • Temperature range: -40°F to 225°F
  • FDA-compliant resin/materials
  • NSF/ANSI/CAN listed
  • Meets USDA and 3-A standards
  • Cut tolerance ±1/4 in