Learn More
Waters Corp Waters Vial, LCMS Certified, Clear Glass, 12 x 32 mm, 2 mL Volume
Catalog No. 50782567 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters LCMS Certified vials are manufactured under tight controls, tested with mass spectrometers, and provided with a certificate of analysis. These vials meet standard dimensional tolerances for autosamplers and feature a PTFE/Silicone septum, ideal for multiple injections and sample storage. Made from chemically inert Type 1, 33-Expansion Borosilicate Glass.
  • Manufactured under tight controls and handling procedures.
  • Mass spectrometer tested; certificate of analysis included.
  • Meets autosampler dimensional tolerances.
  • LCMS certified via unbiased ionized mass tests.
  • PTFE/Silicone septum for multiple injections and storage.
  • Septum working temperature: -40°C to 200°C.
  • Universal screw neck for a tight seal.
  • Constructed from chemically inert Type 1, 33-Expansion Borosilicate Glass.
  • Includes 100 vials per order.
Catalog No. 50-782-567 Supplier Waters Corp Supplier No. 600000751CV
Waters LCMS Certified vials are manufactured under tight controls, tested with mass spectrometers, and provided with a certificate of analysis. These vials meet standard dimensional tolerances for autosamplers and feature a PTFE/Silicone septum, ideal for multiple injections and sample storage. Made from chemically inert Type 1, 33-Expansion Borosilicate Glass.
  • Manufactured under tight controls and handling procedures.
  • Mass spectrometer tested; certificate of analysis included.
  • Meets autosampler dimensional tolerances.
  • LCMS certified via unbiased ionized mass tests.
  • PTFE/Silicone septum for multiple injections and storage.
  • Septum working temperature: -40°C to 200°C.
  • Universal screw neck for a tight seal.
  • Constructed from chemically inert Type 1, 33-Expansion Borosilicate Glass.
  • Includes 100 vials per order.