Learn More
Waters Corp Snap Neck Vial, Polypropylene, 12 x 32 mm, 750 μL
Catalog No. 50791286 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This polypropylene snap neck vial is a cost-effective alternative to glass, suitable for autosamplers and applications where compound sticking to glass is a concern.
  • Clean polypropylene-molded vial.
  • Preferred in applications where there is a concern that compound or molecule may stick to a polar glass surface.
  • Cost-effective alternative to glass.
  • Suitable for autosamplers.
Catalog No. 50-791-286 Supplier Waters Corp Supplier No. 186005224
This polypropylene snap neck vial is a cost-effective alternative to glass, suitable for autosamplers and applications where compound sticking to glass is a concern.
  • Clean polypropylene-molded vial.
  • Preferred in applications where there is a concern that compound or molecule may stick to a polar glass surface.
  • Cost-effective alternative to glass.
  • Suitable for autosamplers.