Learn More
Waters Corp Waters Screw Neck Vial, Polypropylene, 12 x 32 mm, 700 μL
Catalog No. 50791283 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This screw neck vial is designed for precise sample storage in a chemically inert environment. It is certified for LC and LC/MS analysis, ensuring reliable, interference-free results.
  • Features an electron-bonded septum that prevents dislodgement.
  • PTFE/Silicone septum provides outstanding resealing and chemical resistance, with an operating temperature range of -40°C to 200°C.
  • Constructed from polypropylene and chemically inert materials for contamination-free results.
  • Certified for LC and LC/MS analysis, undergoing rigorous testing for cleanliness and low analyte adsorption.
  • Dimensions: 12 x 32 mm, with a volume capacity of 700 μL.
  • Includes vial, cap, and septum, with 100 units per package.
Catalog No. 50-791-283 Supplier Waters Corp Supplier No. 186005221
This screw neck vial is designed for precise sample storage in a chemically inert environment. It is certified for LC and LC/MS analysis, ensuring reliable, interference-free results.
  • Features an electron-bonded septum that prevents dislodgement.
  • PTFE/Silicone septum provides outstanding resealing and chemical resistance, with an operating temperature range of -40°C to 200°C.
  • Constructed from polypropylene and chemically inert materials for contamination-free results.
  • Certified for LC and LC/MS analysis, undergoing rigorous testing for cleanliness and low analyte adsorption.
  • Dimensions: 12 x 32 mm, with a volume capacity of 700 μL.
  • Includes vial, cap, and septum, with 100 units per package.