Learn More
Waters Corp Waters Screw Neck Vial, Polypropylene, 12 x 32 mm, 700 μL
Catalog No. 50466127 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This polypropylene screw neck vial is designed for sample preparation and is suitable for both liquid and solid sample storage. With dimensions of 12 x 32 mm and a 700 μL volume, it is compatible with most laboratory autosamplers. The high-quality polypropylene material offers excellent chemical resistance and a secure screw-neck design, making it suitable for long-term sample storage.
  • Suitable for both liquid and solid sample storage
  • Compatible with most laboratory autosamplers
  • Offers excellent chemical resistance
  • Suitable for long-term sample storage due to secure screw-neck design and high-quality polypropylene material
Catalog No. 50-466-127 Supplier Waters Corp Supplier No. 186005219
This polypropylene screw neck vial is designed for sample preparation and is suitable for both liquid and solid sample storage. With dimensions of 12 x 32 mm and a 700 μL volume, it is compatible with most laboratory autosamplers. The high-quality polypropylene material offers excellent chemical resistance and a secure screw-neck design, making it suitable for long-term sample storage.
  • Suitable for both liquid and solid sample storage
  • Compatible with most laboratory autosamplers
  • Offers excellent chemical resistance
  • Suitable for long-term sample storage due to secure screw-neck design and high-quality polypropylene material