Learn More
Analytical Sales & Services 12 x 32mm, 2mL Volume, Clear Glass Shell Vials and Conical Snap Plugs
Catalog No. 5000804661
Encompass
Advantage™ 12 x 32mm, 2mL Volume, Clear Glass Shell Vials and Conical Snap Plugs\n\nQty/Pkg: 1000 (case)
Catalog No. 50-008-04661 Supplier Analytical Sales & Services Supplier No. 31271CASE
Advantage™ 12 x 32mm, 2mL Volume, Clear Glass Shell Vials and Conical Snap Plugs\n\nQty/Pkg: 1000 (case)