Learn More
Essendant Standard Aluminum Foil Roll, 12"" x 1000 ft, Silver
Catalog No. NC1129620
Encompass_Preferred
Can withstand both hot and cold foods. Made from 98.5% aluminum with the balance made up of iron and silicon, which are added to give greater strength and puncture resistance.
Catalog No. NC1129620 Supplier Essendant Supplier No. RFP611
Can withstand both hot and cold foods. Made from 98.5% aluminum with the balance made up of iron and silicon, which are added to give greater strength and puncture resistance.