Learn More
Cayman Chemical 12 Z 15 Z-HenIcosadIenoIc 10mg
SDP
Catalog No. 502986275
Encompass_Preferred
A very long-chain polyunsaturated fatty acid; a positional isomer of heneicosadienoic acid that is rare in living organisms
Catalog No. 50-298-6275 Supplier Cayman Chemical Supplier No. 2259410 MG
A very long-chain polyunsaturated fatty acid; a positional isomer of heneicosadienoic acid that is rare in living organisms