Learn More
Agilent Technologies 120 EC-C18, 4.6 MM, 4UM, 3/PK
Catalog No. NC2242133
Encompass
InfinityLab Poroshell 120 EC-C18, 4.6 mm, 4 µm, UHPLC guard, 3 per pack
Catalog No. NC2242133 Supplier Agilent Technologies Supplier No. 820750916
InfinityLab Poroshell 120 EC-C18, 4.6 mm, 4 µm, UHPLC guard, 3 per pack