Learn More
MOLDPRO INC 120 mL Titration Beaker, 44.8 x 113 mm, Clear, 275/case, Flat Bottom, PP Homopolymer
Catalog No. NC2233836
Encompass_Preferred
MoldPro MP-129 fits Metrohm Omnis Pick & Place platform titration. Comparable to Metrohm 6.01400.200. Fits Sample Rack 6.02041.030 used with Sample Robots S, M and L. • 275 beakers/case • Made from virgin polypropylene • Excellent visibility • Good resistance to acids Methods: • ASTM D 664, IP 177 • ASTM D 2896 • ISO 3771
This item is not returnable. View return policy
Catalog No. NC2233836 Supplier MOLDPRO INC Supplier No. MP129
MoldPro MP-129 fits Metrohm Omnis Pick & Place platform titration. Comparable to Metrohm 6.01400.200. Fits Sample Rack 6.02041.030 used with Sample Robots S, M and L. • 275 beakers/case • Made from virgin polypropylene • Excellent visibility • Good resistance to acids Methods: • ASTM D 664, IP 177 • ASTM D 2896 • ISO 3771