Learn More
Chemglass Life Sciences Flask Cork Stand, 120 mm OD x 70 mm ID - Fits 500-1,000 mL Round-Bottom Flasks
SDP
Catalog No. NC3037299
Encompass_Preferred
This cork support ring provides a concave, cushioned seat to stabilize round-bottom flasks during setup, heating, or storage. Its cork construction dampens vibration and helps prevent glassware from slipping on benchtop surfaces.
  • Concave recess supports round-bottom flasks securely.
  • Cork construction cushions glassware and resists slipping.
  • 120 mm outside diameter and 70 mm inside diameter for a secure fit.
  • Designed to fit 500-1,000 mL round-bottom flasks.
  • Sold as a single unit for easy replacement or spares.
Catalog No. NC3037299 Supplier Chemglass Life Sciences Supplier No. CG3060120
Add to Cart
Add to Cart
This cork support ring provides a concave, cushioned seat to stabilize round-bottom flasks during setup, heating, or storage. Its cork construction dampens vibration and helps prevent glassware from slipping on benchtop surfaces.
  • Concave recess supports round-bottom flasks securely.
  • Cork construction cushions glassware and resists slipping.
  • 120 mm outside diameter and 70 mm inside diameter for a secure fit.
  • Designed to fit 500-1,000 mL round-bottom flasks.
  • Sold as a single unit for easy replacement or spares.